Anti-ENPP7 Rabbit Recombinant Polyclonal Antibody (Biotin)

Supplier: AFG Bioscience
A58633-100UL A58633-50UL
77830-378EA 521.4 CAD
77830-378 77830-379
Anti-ENPP7 Rabbit Recombinant Polyclonal Antibody (Biotin)
Antibodies
Type:
Antigen: ENPP7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR