Anti-III Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50058-100UG A50058-50UG
77814-576EA 521.4 CAD
77814-576 77814-577
Anti-III Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: III
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Enterobacteria phage M13
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR