Anti-PYCR1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A32544-100UG A32544-50UG
77796-249EA 521.4 CAD
77796-249 77796-250
Anti-PYCR1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: PYCR1
Clonality: Recombinant Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR