Anti-THADA Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A58656-100UG A58656-50UG
77830-423EA 521.4 CAD
77830-423 77830-424
Anti-THADA Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: THADA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR