Anti-BOC Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A67445-100UL A67445-50UL
77836-856EA 521.4 CAD
77836-856 77836-857
Anti-BOC Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: BOC
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR