Anti-HMGB2 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A66747-100UG A66747-50UG
77835-551EA 521.4 CAD
77835-551 77835-552
Anti-HMGB2 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: HMGB2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR