Anti-GATD3A Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A31390-100UG A31390-50UG
77794-415EA 521.4 CAD
77794-415 77794-416
Anti-GATD3A Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GATD3A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR