Anti-PLD3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A31384-100UG A31384-50UG
77794-403EA 521.4 CAD
77794-403 77794-404
Anti-PLD3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200, IF: 1:50 to 1:200

Type:
Antigen: PLD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR