Anti-TIMM8A Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A36606-100UL A36606-50UL
77802-577EA 521.4 CAD
77802-577 77802-578
Anti-TIMM8A Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: TIMM8A
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR