Anti-HECTD3 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A57669-100UG A57669-50UG
77828-557EA 521.4 CAD
77828-557 77828-558
Anti-HECTD3 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: HECTD3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR