Anti-Tpsab1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A37328-100UG A37328-50UG
77811-399EA 521.4 CAD
77811-399 77803-399
Anti-Tpsab1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Tpsab1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR