Anti-KCNJ8 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A54266-100UG A54266-50UG
77822-169EA 521.4 CAD
77822-169 77822-170
Anti-KCNJ8 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:50 to 1:200

Type:
Antigen: KCNJ8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR