Anti-SET Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A68737-100UG A68737-50UG
77839-288EA 521.4 CAD
77839-288 77839-289
Anti-SET Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SET
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR