Anti-OSBPL7 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66651-100UL A66651-50UL
77835-365EA 521.4 CAD
77835-365 77835-366
Anti-OSBPL7 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: OSBPL7
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR