Anti-SLURP1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A35145-100UG A35145-50UG
77800-246EA 521.4 CAD
77800-246 77800-247
Anti-SLURP1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLURP1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR