Anti-NARS2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A65988-100UG A65988-50UG
77834-121EA 521.4 CAD
77834-121 77834-122
Anti-NARS2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: NARS2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR