Anti-Beta-mammal toxin Css4 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51182-100UG A51182-50UG
77816-691EA 521.4 CAD
77816-691 77816-692
Anti-Beta-mammal toxin Css4 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Beta-mammal toxin Css4
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Centruroides suffusus suffusus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR