Anti-SLC6A8 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A40570-50UG A40570-100UG
77807-860EA 355.5 CAD
77807-860 77807-859
Anti-SLC6A8 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: SLC6A8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR