Anti-MEP1A Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A55352-100UL A55352-50UL
77824-212EA 521.4 CAD
77824-212 77824-213
Anti-MEP1A Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: MEP1A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR