Anti-MYO5A Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66127-100UG A66127-50UG
77834-383EA 521.4 CAD
77834-383 77834-384
Anti-MYO5A Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: MYO5A
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR