Anti-cyp26b1 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A57559-100UG A57559-50UG
77828-355EA 521.4 CAD
77828-355 77828-356
Anti-cyp26b1 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: cyp26b1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Zebrafish
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR