Anti-ZMYND11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A55494-100UG A55494-50UG
77824-484EA 521.4 CAD
77824-484 77824-485
Anti-ZMYND11 Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: ZMYND11
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR