Anti-M6PR Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A38591-100UG A38591-50UG
77804-803EA 521.4 CAD
77804-803 77804-804
Anti-M6PR Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: M6PR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR