Anti-CUP1-1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A50280-100UL A50280-50UL
77814-984EA 521.4 CAD
77814-984 77814-985
Anti-CUP1-1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: CUP1-1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Saccharomyces cerevisiae
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR