Anti-AR Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A41404-100UG A41404-50UG
77809-094EA 521.4 CAD
77809-094 77809-095
Anti-AR Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: AR
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR