Anti-Pla2g5 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A31339-100UG A31339-50UG
77794-329EA 521.4 CAD
77794-329 77794-330
Anti-Pla2g5 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: Pla2g5
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR