Anti-ZNF432 Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A38960-100UG A38960-50UG
77805-449EA 521.4 CAD
77805-449 77805-450
Anti-ZNF432 Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: ZNF432
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR