Anti-EHD2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A68560-100UG A68560-50UG
77838-951EA 521.4 CAD
77838-951 77838-952
Anti-EHD2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: EHD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR