Anti-ARHGAP29 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A56684-100UG A56684-50UG
77826-733EA 521.4 CAD
77826-733 77826-734
Anti-ARHGAP29 Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: ARHGAP29
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR