Anti-PTPN3 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A32452-100UG A32452-50UG
77796-104EA 521.4 CAD
77796-104 77796-105
Anti-PTPN3 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:500 to 1:1000, IF: 1:200 to 1:500

Type:
Antigen: PTPN3
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR