Anti-SLC16A8 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A34845-100UG A34845-50UG
77799-762EA 521.4 CAD
77799-762 77799-763
Anti-SLC16A8 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLC16A8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR