Anti-POLD2 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A31563-100UG A31563-50UG
77794-689EA 521.4 CAD
77794-689 77794-690
Anti-POLD2 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:200 to 1:500

Type:
Antigen: POLD2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR