Anti-ACS8 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A67968-100UL A67968-50UL
77837-835EA 521.4 CAD
77837-835 77837-836
Anti-ACS8 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: ACS8
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Arabidopsis thaliana
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR