Anti-POLH Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A31581-100UG A31581-50UG
77794-714EA 521.4 CAD
77794-714 77794-715
Anti-POLH Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: POLH
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR