Anti-RIT2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A66512-100UG A66512-50UG
77835-099EA 521.4 CAD
77835-099 77835-100
Anti-RIT2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: RIT2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR