Anti-GK Rabbit Recombinant Polyclonal Antibody (FITC)

Supplier: AFG Bioscience
A49998-100UG A49998-50UG
77814-466EA 521.4 CAD
77814-466 77814-467
Anti-GK Rabbit Recombinant Polyclonal Antibody (FITC)
Antibodies
Type:
Antigen: GK
Clonality: Recombinant Polyclonal
Clone:
Conjugation: FITC
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR