Anti-CPA2 Rabbit Polyclonal Antibody

Supplier: AFG Bioscience
A53136-100UL A53136-50UL
77820-221EA 521.4 CAD
77820-221 77820-222
Anti-CPA2 Rabbit Polyclonal Antibody
Antibodies
Recommended dilution: ELISA: 1:2000 to 1:5000, WB: 1:500 to 1:2000, IHC: 1:50 to 1:200

Type:
Antigen: CPA2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human;Mouse;Rat
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR