Anti-GPS1 Rabbit Recombinant Antibody

Supplier: AFG Bioscience
A54618-100UL A54618-50UL
77822-838EA 521.4 CAD
77822-838 77822-839
Anti-GPS1 Rabbit Recombinant Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: GPS1
Clonality: Recombinant
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR