Anti-CELSR1 Rabbit Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A67842-100UL A67842-50UL
77837-612EA 521.4 CAD
77837-612 77837-613
Anti-CELSR1 Rabbit Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: CELSR1
Clonality: Polyclonal
Clone:
Conjugation: Biotinylated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR