Anti-L-lactate dehydrogenase Rabbit Recombinant Polyclonal Antibody (Biotinylated)

Supplier: AFG Bioscience
A56075-100UG A56075-50UG
77825-592EA 521.4 CAD
77825-592 77825-593
Anti-L-lactate dehydrogenase Rabbit Recombinant Polyclonal Antibody (Biotinylated)
Antibodies
Type:
Antigen: L-lactate dehydrogenase
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Plasmodium falciparum
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR