Anti-MCIDAS Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A52539-100UG A52539-50UG
77819-143EA 521.4 CAD
77819-143 77819-144
Anti-MCIDAS Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC:1:20 to 1:200

Type:
Antigen: MCIDAS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR