Anti-PTPRN Rabbit Recombinant Antibody (Biotinylated)

Supplier: AFG Bioscience
A54323-100UG A54323-50UG
77822-276EA 521.4 CAD
77822-276 77822-277
Anti-PTPRN Rabbit Recombinant Antibody (Biotinylated)
Antibodies
Type:
Antigen: PTPRN
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR