Anti-HLA-DMA Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A50923-100UG A50923-50UG
77816-205EA 521.4 CAD
77816-205 77816-206
Anti-HLA-DMA Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: HLA-DMA
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR