Anti-NS Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A51294-100UG A51294-50UG
77816-905EA 521.4 CAD
77816-905 77816-906
Anti-NS Rabbit Recombinant Polyclonal Antibody
Antibodies
Type:
Antigen: NS
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Influenza A virus
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR