Anti-Copper-metallothionein Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51093-100UL A51093-50UL
77816-523EA 521.4 CAD
77816-523 77816-524
Anti-Copper-metallothionein Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: Copper-metallothionein
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Roman snail
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR