Anti-KIFC1 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A68077-100UG A68077-50UG
77838-039EA 521.4 CAD
77838-039 77838-040
Anti-KIFC1 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IF: 1:200 to 1:500

Type:
Antigen: KIFC1
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR