Anti-EHD1 Rabbit Recombinant Antibody (Biotinylated)

Supplier: AFG Bioscience
A70197-100UG A70197-50UG
77842-046EA 521.4 CAD
77842-046 77842-047
Anti-EHD1 Rabbit Recombinant Antibody (Biotinylated)
Antibodies
Type:
Antigen: EHD1
Clonality: Recombinant
Clone:
Conjugation: Biotin
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR