Anti-SLC5A2 Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A35067-100UG A35067-50UG
77800-124EA 521.4 CAD
77800-124 77800-125
Anti-SLC5A2 Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: SLC5A2
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR