Anti-X Rabbit Recombinant Polyclonal Antibody (HRP)

Supplier: AFG Bioscience
A51271-100UG A51271-50UG
77816-859EA 521.4 CAD
77816-859 77816-860
Anti-X Rabbit Recombinant Polyclonal Antibody (HRP)
Antibodies
Type:
Antigen: X
Clonality: Recombinant Polyclonal
Clone:
Conjugation: HRP
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Hepatitis B virus genotype D subtype ayw
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR