Anti-KAT14 Rabbit Recombinant Polyclonal Antibody

Supplier: AFG Bioscience
A69501-100UL A69501-50UL
77840-725EA 521.4 CAD
77840-725 77840-726
Anti-KAT14 Rabbit Recombinant Polyclonal Antibody
Antibodies
Recommended dilution: IHC: 1:20 to 1:200

Type:
Antigen: KAT14
Clonality: Recombinant Polyclonal
Clone:
Conjugation: Unconjugated
Epitope:
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR